Assay Details
| Gene Symbol: GCM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008480.2 | NP_001008480.1 | 2 - 3 | 300 | 72 | |
| GenBank mRNA | AB175672.1 | - | 2 - 3 | 301 | 72 | |
More Information
Set Membership: |
|

GCM2
Genomic Map