Assay Details
| Gene Symbol: DYX1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008674.1 | NP_001008674.1 | 1 - 2 | 220 | 82 | |
| GenBank mRNA | AY464141.1 | - | 1 - 2 | 220 | 82 | |
| CR389904.1 | - | 1 - 2 | 198 | 82 | ||
More Information
Set Membership: |
|

DYX1C1
Genomic Map