Assay Details
| Gene Symbol: LEFTY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204764.2 | NP_990095.2 | 1 - 2 | 318 | 96 | |
| XM_015283617.2 | XP_015139103.1 | 2 - 3 | 3112 | 96 | ||
| GenBank mRNA | AB031398.1 | - | 1 - 2 | 317 | 96 | |
| AF179483.1 | - | 1 - 2 | 137 | 96 | ||
More Information
Set Membership: |
|

LEFTY1
Genomic Map