Assay Details
| Gene Symbol: LEFTY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204764.2 | NP_990095.2 | 2 - 3 | 550 | 70 | |
| XM_015283617.2 | XP_015139103.1 | 3 - 4 | 3344 | 70 | ||
| GenBank mRNA | AB031398.1 | - | 2 - 3 | 549 | 70 | |
| AF179483.1 | - | 2 - 3 | 369 | 70 | ||
More Information
Set Membership: |
|

LEFTY1
Genomic Map