Assay Details
| Gene Symbol: CCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204776.1 | NP_990107.1 | 2 - 3 | 252 | 88 | |
| GenBank mRNA | FR874030.1 | - | 2 - 3 | 190 | 88 | |
| L34552.1 | - | 2 - 3 | 252 | 88 | ||
More Information
Set Membership: |
|

CCL1
Genomic Map