Assay Details
| Gene Symbol: ESR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204794.2 | NP_990125.1 | 4 - 5 | 903 | 65 | |
| GenBank mRNA | AB036415.1 | - | 4 - 5 | 907 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESR2| Gene Symbol: ESR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204794.2 | NP_990125.1 | 4 - 5 | 903 | 65 | |
| GenBank mRNA | AB036415.1 | - | 4 - 5 | 907 | 65 | |
Set Membership: |
|