Assay Details
| Gene Symbol: SOCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137648.1 | NP_001131120.1 | 1 | 554 | 93 | |
| XM_015294233.2 | XP_015149719.1 | 2 | 645 | 93 | ||
| GenBank mRNA | BX933215.2 | - | 1 | 471 | 93 | |
| EF554725.1 | - | 1 | 554 | 93 | ||
More Information
Set Membership: |
|

SOCS1
Genomic Map