Assay Details
| Gene Symbol: TLR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204278.1 | NP_989609.1 | 1 - 2 | 106 | 75 | |
| XM_025149461.1 | XP_025005229.1 | 1 - 2 | 120 | 75 | ||
| GenBank mRNA | AB046119.2 | - | 1 - 2 | 106 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_204278.1 transcripts.
Set Membership: |
|

TLR2A
Genomic Map