Assay Details
| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204121.1 | NP_989452.1 | 2 | 1754 | 88 | |
| XM_015298359.2 | XP_015153845.1 | 2 | 2078 | 88 | ||
| GenBank mRNA | AF393850.1 | - | 2 | 1754 | 88 | |
More Information
Set Membership: |
|

NHLH1
Genomic Map