Assay Details
| Gene Symbol: HCRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204185.2 | NP_989516.1 | 2 | 551 | 111 | |
| GenBank mRNA | AB056748.1 | - | 2 | 554 | 111 | |
| CR390067.1 | - | 2 | 549 | 111 | ||
More Information
Set Membership: |
|

HCRT
Genomic Map