Assay Details
| Gene Symbol: FGF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204399.1 | NP_989730.1 | 3 | 648 | 114 | |
| GenBank mRNA | AB108842.1 | - | 3 | 648 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF9| Gene Symbol: FGF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204399.1 | NP_989730.1 | 3 | 648 | 114 | |
| GenBank mRNA | AB108842.1 | - | 3 | 648 | 114 | |
Set Membership: |
|