Assay Details
| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204620.1 | NP_989951.1 | 2 | 745 | 100 | |
| XM_015289379.2 | XP_015144865.1 | 2 | 998 | 100 | ||
| GenBank mRNA | JF317542.1 | - | 2 | 718 | 100 | |
| M81249.1 | - | 2 | 745 | 100 | ||
More Information
Set Membership: |
|

HOXD11
Genomic Map