Assay Details
| Gene Symbol: POU4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204759.1 | NP_990090.1 | 2 | 751 | 108 | |
| GenBank mRNA | X91998.1 | - | 751 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU4F3| Gene Symbol: POU4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204759.1 | NP_990090.1 | 2 | 751 | 108 | |
| GenBank mRNA | X91998.1 | - | 751 | 108 | ||
Set Membership: |
|