Assay Details
| Gene Symbol: SOX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204761.1 | NP_990092.1 | 1 | 751 | 99 | |
| GenBank mRNA | AF193760.2 | - | 1 | 751 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX14| Gene Symbol: SOX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204761.1 | NP_990092.1 | 1 | 751 | 99 | |
| GenBank mRNA | AF193760.2 | - | 1 | 751 | 99 | |
Set Membership: |
|