Assay Details
| Gene Symbol: XPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204853.2 | NP_990184.2 | 6 | 1297 | 72 | |
| XM_025144654.1 | XP_025000422.1 | 7 | 7294 | 72 | ||
| GenBank mRNA | D31896.1 | - | 6 | 1248 | 72 | |
More Information
Set Membership: |
|

XPA
Genomic Map