Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205325.1 | NP_990656.1 | 451 | 69 | ||
| GenBank mRNA | X55998.1 | - | 451 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARF5| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205325.1 | NP_990656.1 | 451 | 69 | ||
| GenBank mRNA | X55998.1 | - | 451 | 69 | ||
Set Membership: |
|