Assay Details
| Gene Symbol: MID1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213575.1 | NP_998740.1 | 1 | 948 | 103 | |
| GenBank mRNA | AY568632.1 | - | 1 | 948 | 103 | |
| BX931309.2 | - | 1 | 890 | 103 | ||
| CR386775.1 | - | 1 | 545 | 103 | ||
More Information
Set Membership: |
|

MID1IP1
Genomic Map