Assay Details
| Gene Symbol: LOC771456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318432.1 | NP_001305361.1 | 5 - 6 | 389 | 78 | |
| XM_004943000.3 | XP_004943057.1 | 4 - 5 | 1407 | 78 | ||
| XM_025152266.1 | XP_025008034.1 | 6 - 7 | 641 | 78 | ||
| XM_025152267.1 | XP_025008035.1 | 6 - 7 | 769 | 78 | ||
| GenBank mRNA | KF271750.1 | - | 5 - 6 | 389 | 78 | |
More Information
Set Membership: |
|

LOC771456
Genomic Map