Assay Details
| Gene Symbol: PPIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166326.1 | NP_001159798.1 | 4 - 5 | 369 | 74 | |
| GenBank mRNA | BX933768.2 | - | 4 - 5 | 422 | 74 | |
| GQ849480.1 | - | 4 - 5 | 369 | 74 | ||
More Information
Set Membership: |
|

PPIA
Genomic Map