Assay Details
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172376.1 | NP_001165847.1 | 1 - 2 | 138 | 79 | |
| GenBank mRNA | GQ149127.1 | - | 1 - 2 | 138 | 79 | |
| KC700625.1 | - | 1 - 2 | 167 | 79 | ||
More Information
Set Membership: |
|

CRNN
Genomic Map