Assay Details
| Gene Symbol: ASCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204412.1 | NP_989743.1 | 1 - 2 | 928 | 74 | |
| GenBank mRNA | L11871.1 | - | 1 - 2 | 774 | 74 | |
| U01339.1 | - | 1 - 2 | 928 | 74 | ||
More Information
Set Membership: |
|

ASCL1
Genomic Map