Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205128.1 | NP_990459.1 | 1 - 2 | 701 | 81 | |
| XM_025144247.1 | XP_025000015.1 | 2 - 3 | 918 | 81 | ||
| XM_025144248.1 | XP_025000016.1 | 2 - 3 | 918 | 81 | ||
| GenBank mRNA | D21837.1 | - | 1 - 2 | 701 | 81 | |
More Information
Set Membership: |
|

OCLN
Genomic Map