Assay Details
| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001744.1 | NP_001001744.1 | 4 - 5 | 836 | 76 | |
| XM_015282323.2 | XP_015137809.1 | 4 - 5 | 1246 | 76 | ||
| XM_015282324.2 | XP_015137810.1 | 4 - 5 | 771 | 76 | ||
| GenBank mRNA | AF091504.1 | - | 4 - 5 | 836 | 76 | |
More Information
Set Membership: |
|

IRX4
Genomic Map