Assay Details
| Gene Symbol: PAQR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008462.1 | NP_001008462.1 | 1 - 2 | 51 | 86 | |
| XM_015285132.2 | XP_015140618.1 | 1 - 2 | 53 | 81 | ||
| GenBank mRNA | AJ720989.1 | - | 1 - 2 | 51 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ720989.1, NM_001008462.1, XM_015285132.2 transcripts.
Set Membership: |
|

PAQR8
Genomic Map