Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008678.1 | NP_001008678.1 | 2 - 3 | 111 | 102 | |
| XM_015273561.2 | XP_015129047.1 | 2 - 3 | 100 | 105 | ||
| XM_015273566.2 | XP_015129052.1 | 2 - 3 | 112 | 98 | ||
| GenBank mRNA | AJ719399.1 | - | 2 - 3 | 111 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ719399.1, NM_001008678.1, XM_015273561.2, XM_015273566.2 transcripts.
Set Membership: |
|

RAB9A
Genomic Map