Assay Details
| Gene Symbol: PTH2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177575.1 | NP_001171046.1 | 1 - 2 | 80 | 59 | |
| GenBank mRNA | DQ914925.1 | - | 1 - 2 | 80 | 59 | |
| FR746109.1 | - | 1 - 2 | 56 | 59 | ||
More Information
Set Membership: |
|

PTH2R
Genomic Map