Assay Details
| Gene Symbol: NDUFV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199171.1 | NP_001186100.1 | 2 - 3 | 226 | 102 | |
| GenBank mRNA | BX950807.1 | - | 2 - 3 | 132 | 102 | |
| HM012718.1 | - | 2 - 3 | 139 | 102 | ||
More Information
Set Membership: |
|

NDUFV3
Genomic Map