Assay Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199230.1 | NP_001186159.1 | 3 - 4 | 324 | 156 | |
| GenBank mRNA | BU466394.1 | - | 3 - 4 | 290 | 157 | |
More Information
Set Membership: |
|

NDUFAF2
Genomic Map