Assay Details
| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199502.1 | NP_001186431.1 | 1 - 2 | 135 | 92 | |
| XM_015283070.2 | XP_015138556.1 | 2 - 3 | 142 | 92 | ||
| XM_015283071.1 | XP_015138557.1 | 1 - 2 | 221 | 92 | ||
| XM_025147135.1 | XP_025002903.1 | 2 - 3 | 230 | 92 | ||
| GenBank mRNA | BI064905.1 | - | 1 - 2 | 136 | 92 | |
More Information
Set Membership: |
|

DEPTOR
Genomic Map