Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204217.2 | NP_001191146.1 | 1 - 2 | 2391 | 68 | |
| NM_001371548.1 | NP_001358477.1 | 8 - 9 | 3146 | 68 | ||
| XM_015280577.2 | XP_015136063.1 | 9 - 10 | 3851 | 68 | ||
| XM_025144612.1 | XP_025000380.1 | 10 - 11 | 3956 | 68 | ||
| XM_025144613.1 | XP_025000381.1 | 10 - 11 | 3956 | 68 | ||
| XM_025144614.1 | XP_025000382.1 | 9 - 10 | 3251 | 68 | ||
| XM_025144616.1 | XP_025000384.1 | 7 - 8 | 3044 | 68 | ||
| GenBank mRNA | BU459818.1 | - | 1 - 2 | 337 | 68 | |
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map