Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204217.2 | NP_001191146.1 | 2 - 3 | 2572 | 69 | |
| XM_025144613.1 | XP_025000381.1 | 11 - 12 | 4137 | 69 | ||
| GenBank mRNA | BU308922.1 | - | 2 - 3 | 561 | 69 | |
| BU459818.1 | - | 2 - 3 | 518 | 69 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map