Assay Details
| Gene Symbol: CENPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204932.1 | NP_001191861.1 | 2 - 3 | 167 | 72 | |
| GenBank mRNA | BX930742.2 | - | 2 - 3 | 167 | 72 | |
| BX933015.2 | - | 2 - 3 | 158 | 72 | ||
More Information
Set Membership: |
|

CENPS
Genomic Map