Assay Details
| Gene Symbol: QRICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257267.1 | NP_001244196.1 | 6 - 7 | 1859 | 67 | |
| XM_015292923.2 | XP_015148409.1 | 6 - 7 | 1916 | 67 | ||
| XM_015292924.2 | XP_015148410.1 | 6 - 7 | 1916 | 67 | ||
| XM_025154394.1 | XP_025010162.1 | 6 - 7 | 2173 | 67 | ||
| XM_025154395.1 | XP_025010163.1 | 7 - 8 | 1953 | 67 | ||
| XM_025154396.1 | XP_025010164.1 | 6 - 7 | 1897 | 67 | ||
| GenBank mRNA | BX935331.2 | - | 4 - 5 | 811 | 67 | |
| CR354106.1 | - | 3 - 4 | 323 | 67 | ||
| CR407006.1 | - | 2 - 3 | 181 | 67 | ||
More Information
Set Membership: |
|

QRICH1
Genomic Map