Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257290.1 | NP_001244219.1 | 3 - 4 | 490 | 63 | |
| GenBank mRNA | BX929493.2 | - | 3 - 4 | 353 | 63 | |
| BX934778.1 | - | 3 - 4 | 490 | 63 | ||
More Information
Set Membership: |
|

TIMMDC1
Genomic Map