Assay Details
| Gene Symbol: ENY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277432.1 | NP_001264361.1 | 2 - 3 | 349 | 87 | |
| NM_001277433.1 | NP_001264362.1 | 3 - 4 | 212 | 87 | ||
| XM_025147116.1 | XP_025002884.1 | 3 - 4 | 277 | 87 | ||
| GenBank mRNA | BU259725.1 | - | 3 - 4 | 206 | 87 | |
| BX258120.3 | - | 2 - 3 | 172 | 87 | ||
| BX272505.4 | - | 2 - 3 | 176 | 87 | ||
| CF252535.1 | - | 3 - 4 | 198 | 87 | ||
| DN930008.1 | - | 2 - 3 | 349 | 87 | ||
More Information
Set Membership: |
|

ENY2
Genomic Map