Assay Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277484.1 | NP_001264413.1 | 1 - 2 | 110 | 68 | |
| XM_015275229.2 | XP_015130715.1 | 2 - 3 | 474 | 68 | ||
| XM_025155430.1 | XP_025011198.1 | 3 - 4 | 666 | 68 | ||
| GenBank mRNA | CN218984.1 | - | 1 - 2 | 110 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277484.1, XM_015275229.2, XM_025155430.1 transcripts.
Set Membership: |
|

HIC2
Genomic Map