Assay Details
| Gene Symbol: CABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277611.1 | NP_001264540.1 | 3 - 4 | 245 | 93 | |
| NR_102411.1 | 3 - 4 | 224 | 93 | |||
| XM_015286223.2 | XP_015141709.2 | 3 - 4 | 697 | 93 | ||
| XM_015286224.2 | XP_015141710.2 | 4 - 5 | 854 | 93 | ||
| XM_025150839.1 | XP_025006607.1 | 3 - 4 | 700 | 93 | ||
| XM_025150840.1 | XP_025006608.1 | 3 - 4 | 712 | 93 | ||
| GenBank mRNA | AJ394204.1 | - | 2 - 3 | 94 | 93 | |
| BX932892.2 | - | 2 - 3 | 176 | 93 | ||
| BX934965.1 | - | 2 - 3 | 235 | 93 | ||
| CR523327.1 | - | 3 - 4 | 245 | 93 | ||
More Information
Set Membership: |
|

CABP2
Genomic Map