Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277637.1 | NP_001264566.1 | 1 - 2 | 186 | 83 | |
| GenBank mRNA | DN929531.1 | - | 1 - 2 | 186 | 83 | |
More Information
Set Membership: |
|

MED9
Genomic Map