Assay Details
| Gene Symbol: ATP5G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277855.1 | NP_001264784.1 | 4 - 5 | 532 | 106 | |
| NM_001277856.1 | NP_001264785.1 | 4 - 5 | 517 | 106 | ||
| NR_102686.1 | 4 - 5 | 573 | 106 | |||
| GenBank mRNA | BU404648.1 | - | 4 - 5 | 575 | 109 | |
| BX931913.1 | - | 5 - 6 | 106 | |||
| CN220040.1 | - | 4 - 5 | 390 | 106 | ||
| CN225005.1 | - | 4 - 5 | 406 | 106 | ||
| CO635700.1 | - | 4 - 5 | 433 | 106 | ||
More Information
Set Membership: |
|

ATP5G3
Genomic Map