Assay Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278020.1 | NP_001264949.1 | 2 - 3 | 170 | 85 | |
GenBank mRNA | BU291350.1 | - | 1 - 2 | 154 | 85 | |
CV856374.1 | - | 1 - 2 | 129 | 85 | ||
DR429670.1 | - | 2 - 3 | 171 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
