Assay Details
| Gene Symbol: TTC30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282395.1 | NP_001269324.1 | 1 | 1824 | 105 | |
| GenBank mRNA | CR733219.1 | - | 1 | 911 | 105 | |
More Information
Set Membership: |
|

TTC30B
Genomic Map