Assay Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287202.1 | NP_001274131.1 | 3 - 4 | 796 | 63 | |
| GenBank mRNA | BU346772.1 | - | 1 - 2 | 286 | 63 | |
More Information
Set Membership: |
|

PHF13
Genomic Map