Assay Details
| Gene Symbol: CDC42SE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302117.1 | NP_001289046.1 | 4 - 5 | 808 | 71 | |
| XM_004949253.3 | XP_004949310.1 | 3 - 4 | 822 | 71 | ||
| XM_004949254.3 | XP_004949311.1 | 2 - 3 | 441 | 71 | ||
| XM_025144760.1 | XP_025000528.1 | 2 - 3 | 379 | 71 | ||
More Information
Set Membership: |
|

CDC42SE2
Genomic Map