Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302139.1 | NP_001289068.1 | 4 - 5 | 515 | 148 | |
| XM_015295640.2 | XP_015151126.1 | 4 - 5 | 653 | 148 | ||
| XM_015295641.2 | XP_015151127.1 | 4 - 5 | 574 | 148 | ||
| XM_015295642.2 | XP_015151128.1 | 4 - 5 | 594 | 148 | ||
| XM_015295643.1 | XP_015151129.1 | 4 - 5 | 389 | 148 | ||
| XM_015295645.2 | XP_015151131.1 | 5 - 6 | 2086 | 148 | ||
| XM_015295646.2 | XP_015151132.1 | 4 - 5 | 3301 | 148 | ||
| XM_015295647.2 | XP_015151133.1 | 4 - 5 | 332 | 148 | ||
| XM_025141875.1 | XP_024997643.1 | 5 - 6 | 719 | 148 | ||
| XM_025141876.1 | XP_024997644.1 | 5 - 6 | 659 | 148 | ||
| XM_025141877.1 | XP_024997645.1 | 4 - 5 | 428 | 148 | ||
| XM_025141878.1 | XP_024997646.1 | 4 - 5 | 653 | 148 | ||
| GenBank mRNA | CR387217.1 | - | 4 - 5 | 515 | 148 | |
More Information
Set Membership: |
|

LYRM9
Genomic Map