Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302173.1 | NP_001289102.1 | 1 - 2 | 161 | 104 | |
| XM_003642629.4 | XP_003642677.1 | 2 - 3 | 263 | 104 | ||
| GenBank mRNA | BX933835.1 | - | 1 - 2 | 161 | 104 | |
| BX934508.1 | - | 1 - 2 | 153 | 104 | ||
More Information
Set Membership: |
|

S100A1
Genomic Map