Assay Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305065.1 | NP_001291994.1 | 1 - 2 | 243 | 107 | |
| GenBank mRNA | GQ504719.1 | - | 1 - 2 | 262 | 107 | |
More Information
Set Membership: |
|

TLCD1
Genomic Map