Assay Details
| Gene Symbol: RBM38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305093.1 | NP_001292022.1 | 3 - 4 | 521 | 133 | |
| GenBank mRNA | AJ720310.1 | - | 3 - 4 | 521 | 133 | |
More Information
Set Membership: |
|

RBM38
Genomic Map