Assay Details
| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305645.1 | NP_001292574.1 | 1 - 2 | 821 | 88 | |
| XM_025147354.1 | XP_025003122.1 | 2 - 3 | 5822 | 88 | ||
| GenBank mRNA | JF923883.1 | - | 1 - 2 | 88 | ||
More Information
Set Membership: |
|

HOXA10
Genomic Map