Assay Details
| Gene Symbol: ELOVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318410.1 | NP_001305339.1 | 3 - 4 | 383 | 109 | |
| GenBank mRNA | HQ621810.1 | - | 3 - 4 | 383 | 109 | |
More Information
Set Membership: |
|

ELOVL3
Genomic Map