Assay Details
| Gene Symbol: SNRPGP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302092.1 | NP_001289021.1 | 4 | 323 | 109 | |
| GenBank mRNA | BU211583.1 | - | 4 | 286 | 109 | |
| DN928912.1 | - | 1 | 89 | 109 | ||
More Information
Set Membership: |
|

SNRPGP15
Genomic Map